Learn More
Fischer Technical Company™ Dehydrated Culture Media and Ingredients
Shop All Fischer Technical Company Products

Description
Successful bacteriological work begins with high-quality media.
- Agar, a solidifying agent, is an essential component of many media
- Available in a variety of formulations and bottle sizes
- For use in preparing plated media, tubed media, bottled media, and slants
Specifications
Specifications
| Form | Dehydrated |
| Format | Bottled |
| Manufacturing Origin | US with Globally Sourced Materials |
| Media Type | Agar Powder |
| Product Type | Microbiology Media |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbiological Tests |
| For Use With (Application) | Microbiological Testing |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.