Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dehydroabietylamine 90.0+%, TCI America™
SDP

Catalog No. D158825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D158825G 25 g
D15885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D158825G Supplier TCI America Supplier No. D158825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1446-61-3
Molecular Formula C20H31N
Molecular Weight (g/mol) 285.475
MDL Number MFCD00213430
InChI Key JVVXZOOGOGPDRZ-SLFFLAALSA-N
Synonym dehydroabietylamine, +-dehydroabietylamine, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine, amine d, dehydroabiethylamine, leelamine, podocarpa-8,11,13-trien-15-amine, 13-isopropyl, 1-phenanthrenemethanamine, 1,2,3,4,4a,9,10,10a-octahydro-1,4a-dimethyl-7-1-methylethyl-, 1r,4as,10ar, pubchem7951, 4-12-00-03005 beilstein handbook reference
PubChem CID 62034
IUPAC Name [(1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl]methanamine
SMILES CC(C)C1=CC2=C(C=C1)C3(CCCC(C3CC2)(C)CN)C

Specifications

Melting Point 40°C
Quantity 25 g
Formula Weight 285.48
Percent Purity ≥90.0% (GC)
Chemical Name or Material (+)-Dehydroabietylamine [Optical Resolving Agent]
EINECSNumber (4)-1242&(7)-0939
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.