Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals

Catalog No. p-2691305
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC364855000

Supplier: Thermo Scientific Chemicals 364855000

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4418-26-2
Molecular Formula C8H7O4Na
MDL Number MFCD00040583
Synonym 3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt

Specifications

Melting Point 295.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Formula Weight 190.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dehydroacetic acid sodium salt, 99%
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.