Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dehydroepiandrosterone Monoclonal Antibody (DHEA(-3):1-19.1 (BGN/117/1191))
GREENER_CHOICE

Catalog No. PIMA174191
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA174191 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA174191 Supplier Invitrogen™ Supplier No. MA174191
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Dehydroepiandrosterone antibody, clone DHEA(-3):1-19.1 detects dehydroepiandrosterone (DHEA) a steroid prohormone which is the precursor of all sex steroids.

Dehydroepiandrosterone (DHEA) is a natural steroid hormone produced from cholesterol by the adrenal glands. DHEA is also produced in the gonads, adipose tissue and the brain. DHEA is structurally similar to, and is a precursor of, androstenedione, testosterone, estradiol, estrone and estrogen. It is the most abundant hormone in the human body. Most of DHEA is sulfated (dehydroepiandrosterone sulfate- DEHAS) before secretion. DHEAS is the sulfated version of DHEA; - this conversion is reversibly catalyzed by sulfotransferase (SULT2A1) primarily in the adrenals, the liver, and small intestines. In blood, most DHEA is found as DHEAS with levels that are about 300 times higher than free DHEA. Blood measurements of DHEAS/DHEA are useful to detect excess adrenal activity as seen in adrenal cancer or hyperplasia, including certain forms of congenital adrenal hyperplasia. Women with polycystic ovary syndrome tend to have normal or mildly elevated levels of DHEAS. [National Center for Biotechnology Information. PubChem Compound Database; CID=5881].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dehydroepiandrosterone
Applications ELISA
Classification Monoclonal
Clone DHEA(-3):1-19.1 (BGN/117/1191)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Androstenolone; C19H28O2; dehydroepiandrosterone sulfate; Dehydroisoandrosterone; DHEA; PRASTERONE
Host Species Mouse
Immunogen DHEA-3-HS-BSA conjugate.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.