Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deionized Water, ACS Reagent, ASTM Type I (D 1193), Reagents

Catalog No. 01336882
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
275 gal.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-882 20 L
01-336-884 10 L
01-336-883 1 L
01-336-881 275 gal.
01-336-880 4 L
01-336-879 500 mL
01-336-878 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-336-882 Supplier Reagents Holdings Llc Supplier No. C234650020F
Add to Cart
Add to Cart

Deionized Water, ACS Reagent, ASTM Type I (D 1193), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Grade ACS
Packaging Cubitainer
Sterility Non-sterile
Quantity 20 L
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Deionized Water, ACS Reagent, ASTM Type I (D 1193)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.