Learn More
Thermo Scientific™ Deionizer Cartridges
Catalog No. 0905052
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Porous Strong Ion Exchange
Strong Anion Exchange
Strong Cation Exchange
Two Bed Ion Exchange
Ultrapure, Oxygen Removal
Ultrapure, Oxygen and Organic Removal
Capacity:
1680 grains
1760 grains
2000 gal.
30 g
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
5000 gal.
5000 gal. (as CaCO3)
785 grains/1000 gal.
875 grains
For Use With (Application):
Anion Removal
Cation Removal
High capacity, two-bed ion exchange
Organic Removal
Oxygen Removal
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure, Oxygen and Organic Removal
Catalog No. 09-050-52
Supplier: Thermo Scientific™ D8809
Only null left
Add to Cart
Add to Cart
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
TRUSTED_SUSTAINABILITY
Specifications
Capacity 455 grains/30 g
Resin Type Ultrapure, Oxygen Removal
For Use With (Application) Ultrapure and Organic Removal
For Use With (Equipment) B-Pure™
Type Deionizer Cartridge
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.