Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEK Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298888AF405
Description
DEK Polyclonal specifically detects DEK in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
DEK | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 405 | |
D6S231EDEK oncogene (DNA binding), DEK oncogene, protein DEK | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human DEK. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
7913 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction