Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEK Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551297

Catalog No. PA551297


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total DEK protein.

DEK is a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DEK
Polyclonal
Unconjugated
DEK
1810019E15Rik; D13H6S231E; D6S231E; DEK; DEK oncogene (DNA binding); DEK proto-oncogene; protein DEK
Rabbit
Antigen affinity chromatography
RUO
7913
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P35659
DEK
Synthetic peptide of human DEK.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.