Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEK Recombinant Rabbit Monoclonal Antibody (JB36-32)
GREENER_CHOICE

Catalog No. PIMA534723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534723 Supplier Invitrogen™ Supplier No. MA534723
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: A431.

DEK is a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEK
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB36-32
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene DEK
Gene Accession No. P35659
Gene Alias 1810019E15Rik; D13H6S231E; D6S231E; DEK; DEK oncogene (DNA binding); DEK proto-oncogene; protein DEK
Gene Symbols DEK
Host Species Rabbit
Immunogen Recombinant protein within Human DEK aa 31-220.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7913
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.