Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Delangin Isoform A Monoclonal Antibody (KT54)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA172534
Description
Delangin Isoform A Monoclonal Antibody for Western Blot, ICC/IF, IP
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
Specifications
Delangin Isoform A | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q6KC79 | |
NIPBL | |
Delangin peptide coupled to BSA. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
KT54 | |
Unconjugated | |
NIPBL | |
4921518A06Rik; 4933421G18Rik; C79399; CDLS; CDLS1; Delangin; delangin homolog; ID; IDN3; IDN3-B; NIPBL; NIPBL, cohesin loading factor; Nipped-B homolog; Nipped-B homolog (Drosophila); nipped-B-like protein; Scc2; SCC2 homolog; sister chromatid cohesion 2 homolog | |
Rat | |
Protein G | |
RUO | |
25836 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction