Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Delangin Isoform B Monoclonal Antibody (KT55)
GREENER_CHOICE

Catalog No. MA172535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA172535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA172535 Supplier Invitrogen™ Supplier No. MA172535
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Delangin Isoform B Monoclonal Antibody for IHC (P)

Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Delangin Isoform B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone KT55
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NIPBL
Gene Accession No. Q6KC79
Gene Alias 4921518A06Rik; 4933421G18Rik; C79399; CDLS; CDLS1; Delangin; delangin homolog; ID; IDN3; IDN3-B; NIPBL; NIPBL, cohesin loading factor; Nipped-B homolog; Nipped-B homolog (Drosophila); nipped-B-like protein; Scc2; SCC2 homolog; sister chromatid cohesion 2 homolog
Gene Symbols NIPBL
Host Species Rat
Immunogen Delangin peptide coupled to BSA.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25836
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.