Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta 2 Catenin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | delta 2 Catenin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
delta 2 Catenin Polyclonal specifically detects delta 2 Catenin in Human samples. It is validated for Western Blot.Specifications
delta 2 Catenin | |
Polyclonal | |
Purified | |
RUO | |
Human | |
catenin (cadherin-associated protein), delta 2 (neural plakophilin-relatedarm-repeat protein), catenin delta-2, Delta-catenin, GT24T-cell delta-catenin, Neural plakophilin-related ARM-repeat protein, Neurojungin, NPRAPneurojungin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 966-1225 of human delta 2 Catenin (NP_001323.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cellular Markers | |
PBS with 50% glycerol, pH7.3. | |
1501 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title