Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ delta Catenin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141117
Description
delta Catenin Polyclonal Antibody for Western Blot, ICC/IF
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
Specifications
| delta Catenin | |
| Polyclonal | |
| Unconjugated | |
| CTNND1 | |
| AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1500, 311163 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O60716 | |
| CTNND1 | |
| Recombinant protein within human delta 1 Catenin/CAS aa 130-320. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction