Alfa Aesar™ delta-Decanolactone, 97%

Manufacturer: Alfa Aesar™ A1175614

Quantity 25g

 View more versions of this product

Catalog No. AAA1175614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 705-86-2
    Molecular Formula C10H18O2
    Formula Weight 170.25
    MDL Number MFCD00006649
    Beilstein 117520
    Synonym delta-Decalactone; 5-Decanolide
    Chemical Name or Material delta-Decanolactone

    Specifications

    Formula Weight 170.25
    Molecular Formula C10H18O2
    MDL Number MFCD00006649
    CAS 705-86-2
    RTECS Number UQ1355000
    TSCA TSCA
    Beilstein 117520
    Synonym delta-Decalactone; 5-Decanolide
    Boiling Point 117° to 120°C (0.02mmHg)
    Melting Point −25°C
    Flash Point 145°C (293°F)
    Quantity 25g
    Chemical Name or Material delta-Decanolactone
    Assay 97%

    Description

    Applications
    Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

    Solubility
    Soluble in water (4 mg/ml at 28°C), alcohols, and propylene glycol.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit