Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta-Hexanolactone, 98%
Click to view available options
Quantity:
25 g
100 g
Description
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 18°C |
Density | 1.04 |
Boiling Point | 231°C |
Flash Point | 105°C (221°F) |
Refractive Index | 1.451 |
Quantity | 25 g |
Beilstein | 80501 |
Solubility Information | Immiscible with water. |
Formula Weight | 114.15 |
Percent Purity | 98% |
Show More |
Safety and Handling
EINECSNumber | 212-511-8 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction