Learn More
Invitrogen™ DLL4 (delta-like 4) Monoclonal Antibody (HMD4-1), Functional Grade, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
Supplier: Invitrogen™ 16594882

Description
Description: This HMD4-1 monoclonal antibody reacts with the extracellular domain of mouse Delta-like 4, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g., Notch 1-4), Delta-like 4 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Delta-like 4 is expressed by thymic lymphoid and stromal cells, as well as macrophages and dendritic cells in the spleen. This molecule is also expressed in non-immune cells, including vascular smooth muscle and endothelial cells. Delta-like 4 plays a role in T cell development, regulation of experimental autoimmune encephalomyelitis, and the formation of mature vasculature. The HMD4-1 monoclonal antibody has been reported to have blocking activity and crossreact to rat. Applications Reported: The HMD4-1 antibody has been reported for use in functional assays. Applications Tested: This HMD4-1 antibody has been tested by flow cytometric analysis of mouse Delta-like 4-transfected cells. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
Specifications
DLL4 (delta-like 4) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
D3ZHH1, Q9JI71 | |
Dll4 | |
Affinity Chromatography | |
RUO | |
311332, 54485 | |
4°C | |
Liquid |
Functional Assay | |
HMD4-1 | |
Functional Grade | |
Dll4 | |
AOS6; delta 4; delta ligand 4; delta like canonical Notch ligand 4; Delta4; delta-like 4 (Drosophila); delta-like 4 homolog; delta-like 4 protein; delta-like protein 4; DLL4; Drosophila Delta homolog 4; hdelta2; MGC126344; notch ligand delta-2; notch ligand DLL4; UNQ1895/PRO4341 | |
Armenian Hamster | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.