Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta-like protein 1 Rat anti-Mouse, Clone: PGPM-1F9, MilliporeSigma™

Catalog No. MABN2284MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2284MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2284MI Supplier MilliporeSigma Supplier No. MABN2284
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Delta-like protein 1 antibody, clone PGPM-1F9 is a highly specific rat monoclonal antibody that targets Delta-like protein 1 and has been tested in Flow Cytometry, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Delta-like protein 1
Applications Flow Cytometry, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone PGPM-1F9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Dll1
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal half of murine Delta-like protein 1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_031891
Test Specificity Clone PGPM-1F9 specifically detects murine Delta Like protein 1. It targets an epitope within 17 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.