Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Delta Np73 Antibody (38C674.2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 38 publications
$206.50 - $499.50
Specifications
Antigen | Delta Np73 |
---|---|
Clone | 38C674.2 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation reported in scientific literature (PMID 19625514), Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen 1:20-1:1000, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224873SS
![]() |
Novus Biologicals
NBP224873SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224873
![]() |
Novus Biologicals
NBP224873 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Delta Np73 Monoclonal specifically detects Delta Np73 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Delta Np73 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
p53-related protein, p73, tumor protein p73 | |
p73 | |
IgG1 κ | |
Protein G purified | |
This antibody is reactive to Human, Mouse and Rat. |
38C674.2 | |
Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation reported in scientific literature (PMID 19625514), Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen 1:20-1:1000, Chromatin Immunoprecipitation (ChIP) | |
Monoclonal | |
Purified | |
RUO | |
O15350 | |
7161 | |
This antibody was developed against a peptide corresponding to a portion of amino acid residues 1-50 of human dNp73. Mouse and human sequences are more than 95 % identical at these amino acid residues. In mouse, alanine at position 8 is replaced by methionine. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title