Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta Opioid R/OPRD1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11079879PE
Description
delta Opioid R/OPRD1 Polyclonal antibody specifically detects delta Opioid R/OPRD1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| delta Opioid R/OPRD1 | |
| Polyclonal | |
| Western Blot | |
| delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD | |
| Synthetic peptide comprising residues 3-17 of the mouse and rat DOR-1 protein. | |
| 0.1 mL | |
| Cancer, GPCR, Membrane Trafficking and Chaperones | |
| 4985 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction