Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Delta Opioid Receptor 3-17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Neuromics™
Rabbit Polyclonal Antibody
Supplier: Neuromics, Inc RA19072100UL

Description
Delta Opioid Receptor 3-17 Polyclonal antibody specifically detects Delta Opioid Receptor 3-17 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western BlotSpecifications
Delta Opioid Receptor 3-17 | |
Polyclonal | |
Unconjugated | |
OPRD1 | |
delta 1 opioid receptor; delta opioid receptor 1; Delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; Opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; OPRD1; Ror-a | |
Synthetic peptide comprising residues 3-17 LVPSARAELQSSPLV of the mouse and rat DOR-1 protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P32300, P33533, P41143 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18386, 24613, 4985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction