Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta-Sarcoglycan Antibody (19G8), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242662C
Description
delta-Sarcoglycan Monoclonal specifically detects delta-Sarcoglycan in Human samples. It is validated for Western Blot, ELISA.Specifications
delta-Sarcoglycan | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SGCD | |
Recombinant human SGCD (57-289aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
19G8 | |
Western Blot, ELISA | |
35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD | |
Mouse | |
Protein A purified | |
RUO | |
6444 | |
Store at 4C in the dark. | |
IgG1b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction