Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta Sarcoglycan (SGCD) Monoclonal Antibody (OTI3D9), TrueMAB™, OriGene
SDP

Catalog No. 502544073
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4073 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4073 Supplier OriGene Supplier No. CF809652
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

delta Sarcoglycan (SGCD) Monoclonal antibody specifically detects delta Sarcoglycan (SGCD) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen delta Sarcoglycan (SGCD)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3D9
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CLP1
Gene Accession No. Q5PQL4, Q92989, Q99LI9
Gene Alias 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kDa; CMD1L; CMD1LSG-delta; DAGD; delta-sarcoglycan; Delta-SG; dystrophin associated glycoprotein, delta sarcoglycan; LGMD2F; MGC22567; placental delta sarcoglycan; sarcoglycan delta; sarcoglycan, delta (35kD dystrophin-associated glycoprotein); sarcoglycan, delta (35kDa dystrophin-associated glycoprotein); sarcoglycan, delta (dystrophin-associated glycoprotein); sarcoglycandelta; sarcoglycan-delta; SGCD; SGCDP; SGD; SGdelta; SG-delta
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-226 of human CLP1 (NP_006822) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10978, 311166, 98985
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.