Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Valerolactam, 98%

Catalog No. p-7082036
Click to view available options
Quantity:
25 g
100 g

CAS: 675-20-7 | C5H9NO | 99.13 g/mol

delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-20-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006037
InChI Key XUWHAWMETYGRKB-UHFFFAOYSA-N
Synonym 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
PubChem CID 12665
ChEBI CHEBI:77761
IUPAC Name piperidin-2-one
SMILES O=C1CCCCN1

Specifications

Melting Point 35°C to 39°C
Boiling Point 255°C to 257°C
Flash Point 147°C (296°F)
Quantity 25 g
Beilstein 106434
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 99.13
Percent Purity ≥98%
Chemical Name or Material delta-Valerolactam
Hazard Category H319-H335
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 211-622-9
RTECSNumber TO0110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.