Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Valerolactam, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
675-20-7
C5H9NO
99.13
MFCD00006037
XUWHAWMETYGRKB-UHFFFAOYSA-N
2-piperidone,2-piperidinone,delta-valerolactam,valerolactim,5-pentanolactam,piperidinone,piperidon,piperidone,alpha-piperidone,piperidone-2
12665
CHEBI:77761
piperidin-2-one
C1CCNC(=O)C1
675-20-7
C5H9NO
99.13
MFCD00006037
XUWHAWMETYGRKB-UHFFFAOYSA-N
2-piperidone,2-piperidinone,delta-valerolactam,valerolactim,5-pentanolactam,piperidinone,piperidon,piperidone,alpha-piperidone,piperidone-2
12665
CHEBI:77761
piperidin-2-one
C1CCNC(=O)C1
Boiling Point 256°C to 262°C
Flash Point 147°C
Viscosity 13.5 mPa.s (50°C)
Specific Gravity 1.073
Melting Point 35°C to 39°C
Packaging Glass bottle
Density 1.0730g/mL
Infrared Spectrum Authentic
Quantity 5g
Color Brown to White
Assay Percent Range 98.5% min. (GC)
Beilstein 21, 238
Solubility Information Solubility in water: 291g/L (25°C). Other solubilities: soluble in most common organic solvents,soluble in ethanol,ether
Formula Weight 99.13
Physical Form Low Melting Crystalline Mass
Percent Purity 99%
Chemical Name or Material δ-Valerolactam, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-622-9

RTECSNumber : TO0110000

TSCA : TSCA