Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deltamethrin , MP Biomedicals™

Catalog No. MP215889825 Shop All MP Biomedicals, Inc Products
Click to view available options
:
25mg

Deltamethrin, Powder

  • This is a Type II pyrethrin and a potent inhibitor of calcineurin (protein phosphatase 2B).
  • More recent study disputes its effectiveness as a calcineurin inhibitor.
  • May also have varied effects on ion channels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7179-49-9
Melting Point 98°C to 101°C
Color Off-white
Molecular Formula C18H37ClN2O7S
Quantity 25 mg
UN Number UN2811 II
Beilstein 6746312
Synonym (S)-α-cyano-3-phenoxybenzyl-(1R)-cis-3-(2,2-dibromvinyl)-2,2-dimethylcyclopropancarboxylate
Solubility Information Soluble in ethanol (25mg/mL) and DMSO (25mg/mL).
InChI Key LFZGYTBWUHCAKF-DCNJEFSFSA-N
SMILES [H+].O.[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name hydrogen (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide hydrate chloride
Molecular Weight (g/mol) 461.01
Vapor Pressure 0.002 mPa (25°C)
Physical Form Powder
Assay 0.983
Show More Show Less
RTECSNumber GZ1233000
storageNote1 Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.