Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deltex 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980247
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0247 Supplier Bioss Supplier No. BS0399R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity.

Specifications

Antigen Deltex 1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Dtx1
Gene Accession No. Q86Y01
Gene Alias deltex 1; deltex 1 homolog (Drosophila); deltex 1, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 1; deltex homolog 1; Deltex1; Dtx1; E3 ubiquitin-protein ligase DTX1; fractionated x-irradiation induced transcript 1; Fxit1; FXI-T1; hDTX1; hDx-1; mDTX1; mKIAA4160; Protein deltex-1; RING-type E3 ubiquitin transferase DTX1
Gene Symbols Dtx1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Deltex-1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1840
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.