Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Demethylasterriquinone B1
GREENER_CHOICESDP

Catalog No. 18195 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-195 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-195 Supplier Tocris Bioscience™ Supplier No. 1819/5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective insulin RTK activator

Selective insulin receptor (IR) activator (EC50 values are 3 - 6 μM for IRTK and 100 μM for IGF1R and EGFR). Increases IR β subunit tyrosine phosphorylation and activation of PI 3-kinase and Akt, but not ERK. Induces glucose uptake in adipocytes and skeletal muscle in vitro, without enhancing vascular proliferation. Binds GAPDH. Also activates Trk by interacting at a site distinct from the neurotrophin-binding site.

Chemical Identifiers

CAS 78860-34-1
Molecular Formula C32H30N2O4

Specifications

Quantity 5 mg
Formula Weight 506.59
Purity 95%
Molecular Weight (g/mol) 506.602
Chemical Name or Material Demethylasterriquinone B1
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.