Learn More
3M™ Demi Fraser Broth Base, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26822 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Volume (Metric) Specimen:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Specimen
S26822 500 g
S26821 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S26822 Supplier 3M™ Supplier No. BP0118500
Add to Cart
Add to Cart
Medium used for the selective enrichment of Listeria species including Listeria monocytogenes
  • 3M™ dehydrated media
  • Convenience of making it when needed
Specifications
Media Type Demi-Fraser Broth Base
Product Type Nutrient Broth
For Use With (Application) Selective enrichment of Listeria species including Listeria monocytogenes
Volume (Metric) Specimen 500 g