Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Demonstration Kit: Chemical Battery
SCI_ED

Catalog No. s96744 Shop All Aldon Products
Click to view available options
:
Chemical Battery
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.

Includes

Materials for five demonstrations: 100 mL Copper (II) Sulfate, 0.5M solution; 200 mL Sodium Sulfate, 0.5M solution; 6 in. Magnesium metal strips UN 1869 (2); 0.75 x 5 in. Copper metal strips (2); 15.9mm dialysis tubing (2); 25 mL Hydrochloric Acid, 3% solution UN 1789; light bulb and wire assembly; empty jar with lid; and instructions

Construct a simple wet-cell battery.

  • When complete, the wet-cell battery will generate 1.5V for 20 to 30 minutes
  • Several cells can be connected in a series to increase the voltage or in parallel to increase the current

Compliance

Package conforms to 49 CFR 173.4.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
Includes 1 x 100 mL Copper (II) Sulfate, 0.5 m solution, 1 x 200 mL Sodium Sulfate, 0.5 m solution, 2 x 6 in. long Magnesium Metal Strips UN1869, 2 x 0.75 x 5 in. Copper Metal Strips, 2 x 15.9 mm dialysis tubing, 1 x 25 mL Hydrochloric Acid, 3% solution UN1789, 1 each light bulb and wire assembly, 1 each empty jar with lid
Quantity 1
Time Range 20 to 30 min.
Dimensions (L x W x H) 6 x 4.25 x 6.875 in.
Class Size 2 Lab Groups
Product Type Chemical Battery Demonstration Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.