Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dendrin Rabbit anti-Mouse, Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB15299I
Description
Anti-Dendrin Antibody is a highly specific rabbit polyclonal antibody that targets Dendrin and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
Dendrin | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This polyclonal antibody recognizes Dendrin in human, mouse, and rat. It targets an epitope of 17 amino acids from the N-terminal region. | |
Stable for 1 year at 2-8°C from date of receipt. |
Immunohistochemistry (Paraffin), Western Blot | |
Please refer to lot specific datasheet. | |
DDN,KIAA0749 | |
KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human Dendrin. | |
100 μg | |
Neuroscience | |
NP_055901 | |
Mouse, Human, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction