Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dendritic Cell Marker Antibody (33D1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP43284646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP43284646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP43284646 Supplier Novus Biologicals Supplier No. NBP143284JF646
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen DendriticCellMarker
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 33D1
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DCs, Dendritic cell
Host Species Rat
Immunogen The immunogen for this antibody was Dendritic Cell Marker.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.