Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dendritic Cell Marker Antibody (33D1), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143284PECY7
Description
Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry.Specifications
Dendritic Cell Marker | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
Rat | |
Protein A or G purified | |
Primary | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
33D1 | |
Flow Cytometry | |
DCs, Dendritic cell | |
The immunogen for this antibody was Dendritic Cell Marker. | |
0.1 mL | |
Mouse | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction