Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dendritic Cell Marker Antibody (AP-MAB0801) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018042 0.025 mg
NBT85474100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018042 Supplier Novus Biologicals Supplier No. NB110854740.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Dendritic Cell Marker
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone AP-MAB0801
Conjugate Unconjugated
Dilution Flow Cytometry 1:100-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:25-1:50, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias DCs, Dendritic cell
Host Species Rat
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse dendritic cells (DCs).
Purification Method Protein G purified
Quantity 0.025 mg
Primary or Secondary Primary
Test Specificity This reacts with Dendritic Cell Inhibitory Receptor 2 on mouse dendritic cells.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.