Learn More
Dendritic Cell Marker Antibody (AP-MAB0801) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Dendritic Cell Marker |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | AP-MAB0801 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:100-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:25-1:50, CyTOF-ready |
| Formulation | A 0.2 um filtered solution in PBS with No Preservative |
| Gene Alias | DCs, Dendritic cell |
| Host Species | Rat |
| Immunogen | This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse dendritic cells (DCs). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.