Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dendritic Cell Marker Antibody (AP-MAB0801), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11085474MFV610
Description
Dendritic Cell Marker Monoclonal antibody specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow CytometrySpecifications
Dendritic Cell Marker | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
AP-MAB0801 | |
Flow Cytometry | |
DCs, Dendritic cell | |
Mouse Dendritic cells | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction