Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus 1 Mouse anti-Virus, Unconjugated, Clone: 271, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Dengue Virus 1 |
---|---|
Clone | 271 |
Applications | ELISA, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF21882MI
|
MilliporeSigma
MABF218825UG |
25 μg |
Each for $182.31
|
|
|||||
MABF21881MI
|
MilliporeSigma
MABF2188100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
Dengue Virus 1 Monoclonal antibody specifically detects Dengue Virus 1 in Virus samples. It is validated for ELISA, Flow Cytometry.Specifications
Dengue Virus 1 | |
ELISA, Flow Cytometry | |
Unconjugated | |
Mouse | |
Virus | |
DENV NS1 protein | |
IgG2b κ | |
Protein G Purified | |
Clone 271 is a mouse monoclonal antibody that detects Dengue virus NS1 protein. Also, displays reactivity with Dengue virus 3 NS1 protein. However, when paired with Dengue virus 1 NS 1 specific antibody (Clone 912), such combination only recognizes Dengue virus 1 NS1 protein and not Dengue virus 3 NS1 protein. It targets an epitope within 16 amino acids from the wing domain. |
271 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_059433 | |
Purified recombinant Dengue virus 1 NS1 protein expressed in mammalian cells. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼48 kDa calculated for NS1 protein; 378.71 kDa for the entire genome polyprotein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title