Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue virus 1 NS1 Human anti-Virus, Clone: OB4.C11.D10.B8, Novus Biologicals™

Human Monoclonal Antibody
$779.50 - $2180.00
Specifications
| Antigen | Dengue virus 1 NS1 |
|---|---|
| Clone | OB4.C11.D10.B8 |
| Concentration | 1 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
Description
Dengue virus 1 NS1 Monoclonal antibody specifically detects Dengue virus 1 NS1 in Virus samples. It is validated for ELISASpecifications
| Dengue virus 1 NS1 | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Human | |
| Virus | |
| Recombinant Dengue virus NS1, a mixture of serotypes 1-4 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| OB4.C11.D10.B8 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS pH 7.4 | |
| IgM | |
| Column Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title