Learn More
Dengue Virus 3/4 NS1 protein Mouse anti-Virus, Clone: 55, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dengue Virus 3/4 NS1 protein |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 55 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | Purified recombinant Dengue virus NS1 protein expressed in mammalian cells. |
| Purification Method | Protein G purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.