Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus 3 NS1 Mouse anti-Virus, Unconjugated, Clone: 411, MilliporeSigma™

Catalog No. MABF21921MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF21921MI 100 μL
MABF21922MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF21921MI Supplier MilliporeSigma Supplier No. MABF2192100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dengue Virus 3 NS1 Monoclonal antibody specifically detects Dengue Virus 3 NS1 in Virus samples. It is validated for ELISA, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 3 NS1
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 411
Conjugate Unconjugated
Gene Alias Denv3 NS1 protein
Host Species Mouse
Immunogen Purified recombinant Dengue virus 3 NS1 protein expressed in mammalian cells.
Molecular Weight of Antigen ∼48 kDa calculated for NS1 protein; 378.71 kDa for the entire genome polyprotein.
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 411 is a mouse monoclonal antibody that specifically detects D3engue virus 3 NS1 protein. It targets an epitope within 17 amino acids from the wing and finger regions.
Target Species Virus
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.