Learn More
Dengue Virus 3 NS1 Mouse anti-Virus, Unconjugated, Clone: 411, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Dengue Virus 3 NS1 |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 411 |
| Conjugate | Unconjugated |
| Gene Alias | Denv3 NS1 protein |
| Host Species | Mouse |
| Immunogen | Purified recombinant Dengue virus 3 NS1 protein expressed in mammalian cells. |
| Molecular Weight of Antigen | ∼48 kDa calculated for NS1 protein; 378.71 kDa for the entire genome polyprotein. |
| Purification Method | Protein G Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.