Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus Antibody (Dengue 1-11(3)), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065685C
Description
Dengue Virus Monoclonal specifically detects Dengue Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Dengue Virus | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
Dengue 1-11(3) | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
DEN 1, DEN1, Dengue hemorrhagic fever, Dengue type 1 virus, DENV 1, DENV1 | |
Made to Viral DENGUE VIRUS | |
0.125 mL | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
NB100-65685 recognizes all four dengue virus serotypes (DEN-1, DEN-2, DEN-3 and DEN-4 least strongly) of the genus Flavivirus. NB100-65685 detects an anti-E/envelope specific band of 61kDa under reducing conditions in Western blotting. A weak secondary band of 80kDa may also be apparent. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction