Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus Capsid Protein Antibody (05), Alexa Fluor™ 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306425AF488
Description
Dengue Virus Capsid Protein Monoclonal antibody specifically detects Dengue Virus Capsid Protein in Virus samples. It is validated for Western Blot, ELISASpecifications
Dengue Virus Capsid Protein | |
Monoclonal | |
Alexa Fluor 488 | |
polyprotein gene | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue virus DENV-2 (Strain New Guinea C) Capsid protein/ DENV-C (AHF52833.1; Met1-Arg100). | |
0.1 mL | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
1494449 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
05 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction