Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus Capsid Protein Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP242822

Catalog No. NBP242822


Only null left
Add to Cart

Description

Description

Specifically detects Dengue Virus Capsid Protein in Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting

Dengue Virus Capsid Protein Polyclonal specifically detects Dengue Virus Capsid Protein in Viral samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Dengue Virus Capsid Protein
Polyclonal
Western Blot 1:500-1:3000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000
POLY
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Dengue virus Capsid protein (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Though raised against DEN2 protein, this antibody is predicted to cross-react with the other serotypes based on comparative sequence analysis. Experimental evidence from an independent lab showed that this antibody does not detect the target protein by western blot in DEN1-infected C6/36 mosquito cell lysates.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.025% Proclin 300
Rabbit
12 kDa
0.1 mL
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
1494449
Virus
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only