Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus NS1 protein Antibody (12), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306437MFV610
Description
Dengue Virus NS1 protein Monoclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for ELISASpecifications
Dengue Virus NS1 protein | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352). | |
0.1 mL | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
Virus | |
Purified |
ELISA | |
12 | |
ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction