Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue virus NS1 types 1, 2, 3 Mouse anti-Virus, Unconjugated, Clone: HM513, Abnova™

Catalog No. 89339284 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-284 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-284 Supplier Abnova Corporation Supplier No. MAB21898
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Dengue virus NS1 types 1, 2, 3 Monoclonal antibody specifically detects Dengue virus NS1 types 1, 2, 3 in Viruses samples. It is validated for LFIA.

Specifications

Antigen Dengue virus NS1 types 1, 2, 3
Applications Lateral Flow
Classification Monoclonal
Clone HM513
Conjugate Unconjugated
Host Species Mouse
Immunogen Native Dengue antigen
Quantity 1 mg
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.