Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus NS2B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP242891

Catalog No. NBP242891


Only null left
Add to Cart

Description

Description

Specifically detects NS2B in Human, Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting

Dengue Virus NS2B Polyclonal specifically detects Dengue Virus NS2B in Viral samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Dengue Virus NS2B
Polyclonal
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:2000
Rabbit
Affinity Purified
RUO
Primary
Virus
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Full length Dengue virus 2 NS2B Recombinant protein (Dengue virus 2 (strain 16681 PDK 53))
0.1 mL
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
1494449
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only