Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus NS4B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP242895
Description
Specifically detects NS4B in Virus samples, and it is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blotting
Dengue Virus NS4B Polyclonal specifically detects Dengue Virus NS4B in Viral samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Dengue Virus NS4B | |
Polyclonal | |
Western Blot 1:1000-1:10000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:2000 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Virus | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Dengue virus NS4B protein (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary. | |
0.1 mL | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
1494449 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction