Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus NS5 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306447MFV500
Description
Dengue Virus NS5 Polyclonal antibody specifically detects Dengue Virus NS5 in Virus samples. It is validated for Western Blot, ELISASpecifications
| Dengue Virus NS5 | |
| Polyclonal | |
| Western Blot, ELISA | |
| Dengue NS5, Dengue NS5 protein | |
| Produced in rabbits immunized with purified, recombinant Dengue virus DENV-2 (Accession#: AAC59275.1; Gly2492-Gln2787). | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 1489713 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction