Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dengue Virus Type 1-3 NS1 Monoclonal Antibody (5169)
GREENER_CHOICE

Catalog No. PIMA533249
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533249 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533249 Supplier Invitrogen™ Supplier No. MA533249
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dengue Virus Type 1-3 NS1 Monoclonal Antibody for ICC/IF, ELISA, LF

Dengue virus infections are caused by four closely related viruses named DEN-1, DEN-2, DEN-3, and DEN-4. These four viruses are called serotypes because each has different interactions with the antibodies in human blood serum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus Type 1-3 NS1
Applications ELISA, Lateral Flow, Immunocytochemistry
Classification Monoclonal
Clone 5169
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Dengue Virus 1; Dengue Virus 2; Dengue Virus 3; Dengue Virus Type 1; Dengue Virus Type 2; Dengue Virus Type 3; DENV; DENV1; DENV2; DENV3
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.