Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dengue Virus Type 2/4 Envelope Recombinant Rabbit Monoclonal Antibody (HL2089)
GREENER_CHOICE

Catalog No. FEMA550377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550377 Supplier Invitrogen™ Supplier No. MA550377
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Dengue Virus Type 2/4 Envelope Recombinant Monoclonal Antibody for Western Blot, ICC/IF

The Dengue Virus Type 2/4 Envelope gene is a target for research and development of vaccines and therapies for the Dengue virus. Dengue fever is a mosquito-borne viral disease that is prevalent in tropical and subtropical regions around the world. The virus has four serotypes, and infection with one serotype does not provide immunity against the others. The Envelope gene is responsible for encoding the structural protein that forms the outer shell of the virus, which plays a crucial role in the virus's entry into host cells and subsequent replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus Type 2/4 Envelope
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2089
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias DENV Type 2/3 Envelope
Host Species Rabbit
Immunogen Recombinant fragment of Dengue virus Envelope protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.